Brilliant Crocein
Brilliant Crocein
Brilliant Crocein
Thermo Scientific Chemicals
Brilliant Crocein
CAS: 5413-75-2 | C22H16N4Na2O7S2 | 558.491 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14642.1850 g
Catalog number ALFA14642.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBrilliant Crocein
CAS5413-75-2
Recommended StorageAmbient temperatures
EINECS Number226-502-1
TSCAYes
View more
Brilliant Crocein is used mainly as a wool, silk, leather paper and leather dyeing. Can also be used for plastic, wood, spices, electrical-controlled aluminum, medicine color. Can make the ink, the heavy metal salt can be used as pigment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Brilliant Crocein is used mainly as a wool, silk, leather paper and leather dyeing. Can also be used for plastic, wood, spices, electrical-controlled aluminum, medicine color. Can make the ink, the heavy metal salt can be used as pigment.
Solubility
Soluble in water
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. JK Pal.;D Godbole.; K Sharma. Staining of proteins on SDS polyacrylamide gels and on nitrocellulose membranes by Alta, a color used as a cosmetic. Journal of Biochemical and Biophysical Methods.2004, 61 (3), 339-347.
  2. WH Cheung.; YS Szeto.; G McKay. Intraparticle diffusion processes during acid dye adsorption onto chitosan. Bioresource Technology.2007, 98 (15), 2897-2904.