2,5-Dihydroxy-1,4-dithiane, 96%
2,5-Dihydroxy-1,4-dithiane, 96%
2,5-Dihydroxy-1,4-dithiane, 96%
2,5-Dihydroxy-1,4-dithiane, 96%
Thermo Scientific Chemicals
2,5-Dihydroxy-1,4-dithiane, 96%
CAS: 40018-26-6 | C4H8O2S2 | 152.226 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14620.1850 g
Catalog number ALFA14620.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material2,5-Dihydroxy-1,4-dithiane
CAS40018-26-6
Health Hazard 1H301-H317-H318
Health Hazard 2GHS H Statement
H318-H302-H317
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Health Hazard 3P261-P264b-P270-P272-P280-P301+P310-P302+P352-P305+P351+P338-P310-P330-P333+P313-P363-P501c