2,3-Diaminopyridine, 98%
2,3-Diaminopyridine, 98%
2,3-Diaminopyridine, 98%
2,3-Diaminopyridine, 98%
Thermo Scientific Chemicals
2,3-Diaminopyridine, 98%
CAS: 452-58-4 | C5H7N3 | 109.13 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14618.065 g
Catalog number ALFA14618.06
Price (MYR)
497.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
497.00
EA
Add to cart
Specifications
Chemical Name or Material2,3-Diaminopyridine
CAS452-58-4
Health Hazard 1H301-H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H300-H311-H331-H315-H319-H335
Fatal if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
2,3-Diaminopyridine was used in the synthesis of 2,3-diaminopyridinium-4-hydroxybenzoate1. It was used as internal standard in analysis of poly(amidoamine)dendrimers having different terminal functionalities by capillary electrophoresis. It was used as reagent in synthesis of imidazopyridines and organometallic complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3-Diaminopyridine was used in the synthesis of 2,3-diaminopyridinium-4-hydroxybenzoate1. It was used as internal standard in analysis of poly(amidoamine)dendrimers having different terminal functionalities by capillary electrophoresis. It was used as reagent in synthesis of imidazopyridines and organometallic complexes.
Solubility
Soluble in water (100 g/L).
Notes
It is sensitive to air. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Hoong-Kun Fun; Kasthuri Balasubramani. 2,3-Diamino-pyridinium 4-hydroxy-benzoate. Acta Crystallographica Section E. 2009, 65, (Pt 7),o1496-o1497
  2. Xiangyang Shi; Istvan Banyai; Wojciech G.Lesniak; Mohammad T.Islam; Istvan Orszagh; Peter Balogh; James R.Baker; Lajos P.Balogh. Capillary electrophoresis of polycationic poly(amidoamine) dendrimers. Electrophoresis. 2005, 26, (15),2949-2959