D-(+)-Cellobiose, 98+%
D-(+)-Cellobiose, 98+%
D-(+)-Cellobiose, 98+%
D-(+)-Cellobiose, 98+%
Thermo Scientific Chemicals
D-(+)-Cellobiose, 98+%
CAS: 528-50-7 | C12H22O11 | 342.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFA14553.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialD-(+)-Cellobiose
CAS528-50-7
Melting Point234°C (decomposition)
Recommended StorageAmbient temperatures
EINECS Number208-436-5
View more
D-(+)-Cellobiose is used to differentiate bacteria based on carbohydrate fermentation abilities. It is a β-glucosidase substrate. Also used as as a chemical raw material and for foods, as a low-sweetness sugar.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-(+)-Cellobiose is used to differentiate bacteria based on carbohydrate fermentation abilities. It is a β-glucosidase substrate. Also used as as a chemical raw material and for foods, as a low-sweetness sugar.
Solubility
Soluble in water (1 g/8ml). Insoluble in ether and ethanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Christopher W.Higham; Duncan Gordon-Smith, Christopher E.Dempsey; Paul M.Wood. Direct 1H NMR evidence for conversion of β-d-cellobiose to cellobionolactone by cellobiose dehydrogenase from Phanerochaete chrysosporium. FEBS Letters. year of publication , 351, (1),128-132
  2. R.Maleszka; P.Y.Wang; Henry Schneider. Yeasts that ferment D-cellobiose as well as D-xylose. Biotechnology Letters. 1982, 4, (2),133-136