2-Bromobenzoyl chloride, 98%
2-Bromobenzoyl chloride, 98%
2-Bromobenzoyl chloride, 98%
2-Bromobenzoyl chloride, 98%
Thermo Scientific Chemicals
2-Bromobenzoyl chloride, 98%
CAS: 7154-66-7 | C7H4BrClO | 219.462 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14520.0910 g
Catalog number ALFA14520.09
Price (MYR)
325.00
EA
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
325.00
EA
Add to cart
Specifications
Chemical Name or Material2-Bromobenzoyl chloride
CAS7154-66-7
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c