Silicon carbide powder, coarse, 46 grit
Silicon carbide powder, coarse, 46 grit
Silicon carbide powder, coarse, 46 grit
Silicon carbide powder, coarse, 46 grit
Silicon carbide powder, coarse, 46 grit
Thermo Scientific Chemicals
Silicon carbide powder, coarse, 46 grit
CAS: 409-21-2 | CSi | 40.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
Catalog number ALFA14470.36
Price (MYR)
551.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
551.00
EA
Add to cart
Specifications
Chemical Name or MaterialSilicon carbide powder
Name Notecoarse, 46 grit
CAS409-21-2
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Silicon carbide is used in abrasives, in polishing and grinding. It is widely used in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are as light emitting diodes and sensors. It is utilized for the production of fertile, transgenic maize plants. It compositely plays an important role in the crack-healing behavior.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Silicon carbide is used in abrasives, in polishing and grinding. It is widely used in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are as light emitting diodes and sensors. It is utilized for the production of fertile, transgenic maize plants. It compositely plays an important role in the crack-healing behavior.
Solubility
Soluble in molten sodium hydroxide, potassium hydroxide and in molten iron.
Notes
Keep it away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Cheng, G.; Chang, T. H.; Qin, Q.; Huang, H.; Zhu, Y. Mechanical Properties of Silicon Carbide Nanowires: Effect of Size-Dependent Defect Density. Electrochem. Commun. 2014, 14 (2), 754-758.
  2. Zhang, X.; Xu, L.; Du, S.; Han, W.; Han, J. Crack-healing behavior of zirconium diboride composite reinforced with silicon carbide whiskers. Scripta Materialia. 2008, 59 (11), 1222-1225.