trans-3-Hydroxycinnamic acid, 99%
trans-3-Hydroxycinnamic acid, 99%
trans-3-Hydroxycinnamic acid, 99%
trans-3-Hydroxycinnamic acid, 99%
Thermo Scientific Chemicals
trans-3-Hydroxycinnamic acid, 99%
CAS: 14755-02-3 | C9H8O3 | 164.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
50 g
Catalog number ALFA14466.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Materialtrans-3-Hydroxycinnamic acid
CAS14755-02-3
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c