Aniline, 99+%
Aniline, 99+%
Aniline, 99+%
Thermo Scientific Chemicals
Aniline, 99+%
CAS: 62-53-3 | C6H7N | 93.13 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14443.22100 g
Catalog number ALFA14443.22
Price (MYR)
286.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
286.00
EA
Add to cart
Specifications
Chemical Name or MaterialAniline
CAS62-53-3
Health Hazard 1H227-H301+H311+H331-H317-H318-H336-H341-H351-H372
Health Hazard 2GHS H Statement
H301-H311-H331-H372-H341-H351-H318-H227-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of causing cancer.
Causes serious eye damage.
Combustible liquid.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P210-P235-P260-P264b-P270-P271-P272-P280g-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P311-P312-P330-P333+P313-P361-P363-P370+P378q-P501c
View more
Aniline is used as a solvent. It has been used as an antiknock compound for gasolines. It has a major application in the preparation of polyurethane which is used in making plastic, building thermal foam insulations and in refrigerators. It is widely used in the preparation of methylene dianiline and related compounds by reacting with formaldehyde. It combines with nitric acid and is utilized as rocket fuel for small missiles and jet assisted take-off. It acts as an additive to rubber and its derivatives such as phenylenediamines and diphenylamine are used as antioxidants. It finds application in dyes and pigments in agricultural and polymer industries. It is employed at a smaller scale in the production of the intrinsically conducting polymer polyaniline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Aniline is used as a solvent. It has been used as an antiknock compound for gasolines. It has a major application in the preparation of polyurethane which is used in making plastic, building thermal foam insulations and in refrigerators. It is widely used in the preparation of methylene dianiline and related compounds by reacting with formaldehyde. It combines with nitric acid and is utilized as rocket fuel for small missiles and jet assisted take-off. It acts as an additive to rubber and its derivatives such as phenylenediamines and diphenylamine are used as antioxidants. It finds application in dyes and pigments in agricultural and polymer industries. It is employed at a smaller scale in the production of the intrinsically conducting polymer polyaniline.
Solubility
Miscible with water.
Notes
Light sensitive. Incompatible with oxidizing agents, iron and iron salts.
RUO – Research Use Only
General References:
  1. Qadir, M. I.; Scholten, J. D.; Dupont, J. Ionic liquid effect: selective aniline oxidative coupling to azoxybenzene by TiO2. Catal. Sci. Technol. 2015, 5 (3), 1459-1462.
  2. Fu, L.; Cai, W.; Wang, A.; Zheng, Y. Photocatalytic hydrogenation of nitrobenzene to aniline over tungsten oxide-silver nanowires. Mater. Lett. 2015, 142, 201-203.