Neopentyl alcohol, 99%
Neopentyl alcohol, 99%
Neopentyl alcohol, 99%
Neopentyl alcohol, 99%
Thermo Scientific Chemicals
Neopentyl alcohol, 99%
CAS: 75-84-3 | C5H12O | 88.15 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14390.1425 g
Catalog number ALFA14390.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialNeopentyl alcohol
CAS75-84-3
Health Hazard 1H228
Health Hazard 2GHS H Statement
H228
Flammable solid.
Health Hazard 3P210-P240-P241-P280-P370+P378q
View more
It is used in the Mitsunobu reaction of methyl salicylate coupling of sterically hindered substrates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the Mitsunobu reaction of methyl salicylate coupling of sterically hindered substrates.
Solubility
Soluble in water at (20°C 40 g/L), alcohol, and ether. Solubility in methanol is almost transparent.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Sudhakar Bhandaru, et al. Synthesis of a C14',15' dihydro derivative of the south hexacyclic steroid unit of cephalostatin 1. Part I : regiospecific Rh[II]-mediated intermolecular oxygen alkylation of a primary neopentyl alcohol.Tetrahedron Lett.,1995,36(46), 8347-8350.
  2. J. D. Slagle, et al. Mechanism of the triphenylphosphine-tetrachloromethane-alcohol reaction: pericyclic or clustered ion pairs?J. Org. Chem.,1981,46(17), 3526-3530.
  3. A method has been described for conversion to the iodide using a combination of methyl iodide and triphenyl phosphite: Org. Synth. Coll., 6, 830 (1988).