1,3-Difluorobenzene, 99%
1,3-Difluorobenzene, 99%
1,3-Difluorobenzene, 99%
1,3-Difluorobenzene, 99%
Thermo Scientific Chemicals
1,3-Difluorobenzene, 99%
CAS: 372-18-9 | C6H4F2 | 114.10 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14387.0910 g
Catalog number ALFA14387.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or Material1,3-Difluorobenzene
CAS372-18-9
Health Hazard 1H225-H332
Health Hazard 2GHS H Statement
H225-H319
Highly flammable liquid and vapour.
Causes serious eye irritation.
Health Hazard 3P210-P233-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
View more
It is used in the laser induced fluorencence matrix study of 1,3-difluorobenzene radical cation. It is also used synthesis of 1,2,3-triazole containing fluconazole analogues.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the laser induced fluorencence matrix study of 1,3-difluorobenzene radical cation. It is also used synthesis of 1,2,3-triazole containing fluconazole analogues.
Solubility
Soluble in water at 20°C 1.1g/L.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. M. T. Kirchner, et. al. 1,3-Difluoro­benzene.Acta Cryst.,2009,E652668-2669.
  2. Lise Nygaard, et al.Microwave spectra and dipole moments of 1,2-difluorobenzene and 1,3-difluorobenzene.Spectrochim. Acta, Part A,1967,23(11), 2813-2819.
  3. Lithiation occurs at the 2-position. For a study of lithiation under various conditions, see: J. Chem. Soc., Perkin 1, 2729 (1995).