N-Phenylanthranilic acid, 99%
N-Phenylanthranilic acid, 99%
N-Phenylanthranilic acid, 99%
N-Phenylanthranilic acid, 99%
Thermo Scientific Chemicals
N-Phenylanthranilic acid, 99%
CAS: 91-40-7 | C13H11NO2 | 213.236 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14340.1425 g
Catalog number ALFA14340.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN-Phenylanthranilic acid
CAS91-40-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
DPC is a potent, non-specific blocker of Cl- channel and chloride-bicarbonate exchange. DPC also inhibits CFTR (cystic fibrosis transmembrane regulator) ATPase activity. It is used to detect vanadium in steel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DPC is a potent, non-specific blocker of Cl- channel and chloride-bicarbonate exchange. DPC also inhibits CFTR (cystic fibrosis transmembrane regulator) ATPase activity. It is used to detect vanadium in steel.
Solubility
Soluble in ethanol, DMSO (50 mg/ml), hot alcohol, and methanol. Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Maite L. Docampo Palaciosa and Rolando F. Pellón Comdom. Synthesis of N-Phenylanthranilic Acid Derivatives Using Water as Solvent in the Presence of Ultrasound Irradiation.Synth. Commun.,2003,33(10), 1771-1775.
  2. Ali Almasirad, et al. Synthesis and Analgesic Activity of 2-Phenoxybenzoic Acid and N-Phenylanthranilic Acid Hydrazides.Biol. Pharm. Bull.,2006,29(6), 1180-1185.