2',4',6'-Trimethoxyacetophenone, 98%
2',4',6'-Trimethoxyacetophenone, 98%
2',4',6'-Trimethoxyacetophenone, 98%
2',4',6'-Trimethoxyacetophenone, 98%
Thermo Scientific Chemicals
2',4',6'-Trimethoxyacetophenone, 98%
CAS: 832-58-6 | C11H14O4 | 210.23 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14328.065 g
Catalog number ALFA14328.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2',4',6'-Trimethoxyacetophenone
CAS832-58-6
Melting Point101°C to 104°C
Recommended StorageAmbient temperatures
TSCANo
View more
2',4',6'-Trimethoxyacetophenone is used in the production of 4,2',4',6'-tetramethoxy-chalcone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2′,4′,6′-Trimethoxyacetophenone is used in the production of 4,2′,4′,6′-tetramethoxy-chalcone.
Solubility
Soluble in chloroform.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Zhang, J.; Liu, M.; Cui, W.; Yang, J.; Yang, B. Total synthesis of luteolin. J. Chem. Res. 2014, 38 (1), 60-61.
  2. Nasir Abbas Bukhari, S.; Jantan, I.; Jasamai, M. Anti-Inflammatory Trends of 1, 3-Diphenyl-2-propen-1-one Derivatives. Mini Rev Med Chem 2013, 13 (1), 87-94.