1,2,4,5-Tetrachlorobenzene, 98%
1,2,4,5-Tetrachlorobenzene, 98%
1,2,4,5-Tetrachlorobenzene, 98%
1,2,4,5-Tetrachlorobenzene, 98%
Thermo Scientific Chemicals
1,2,4,5-Tetrachlorobenzene, 98%
CAS: 95-94-3 | C6H2Cl4 | 215.882 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14318.0910 g
Catalog number ALFA14318.09
Price (MYR)
286.00
EA
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
286.00
EA
Add to cart
Specifications
Chemical Name or Material1,2,4,5-Tetrachlorobenzene
Melting Point138°C to 142°C
CAS95-94-3
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
View more
It finds its application as a high melting diluent in the eutectic crystallization of pseudo binary systems of polyethylene and high melting diluents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds its application as a high melting diluent in the eutectic crystallization of pseudo binary systems of polyethylene and high melting diluents.
Solubility
Insoluble in water. Solubility in toluene is almost transparent.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Peter Sander, et.al. Degradation of 1,2,4-Trichloro- and 1,2,4,5-Tetrachlorobenzene by Pseudomonas Strains.Appl. Environ. Microbiol.,1991,57(5), 1430-1440.
  2. Stefan Beil, et al. Genetic and Biochemical Characterization of the Broad Spectrum Chlorobenzene Dioxygenase from Burkholderia Sp. Strain PS12 — Dechlorination of 1,2,4,5-Tetrachlorobenzene.Eur. J. Biochem.,1997,247(1), 190-199.