3-Nitrobenzyl bromide, 98+%
3-Nitrobenzyl bromide, 98+%
3-Nitrobenzyl bromide, 98+%
3-Nitrobenzyl bromide, 98+%
Thermo Scientific Chemicals
3-Nitrobenzyl bromide, 98+%
CAS: 3958-57-4 | C7H6BrNO2 | 216.034 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14310.065 g
Catalog number ALFA14310.06
Price (MYR)
365.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
365.00
EA
Add to cart
Specifications
Chemical Name or Material3-Nitrobenzyl bromide
CAS3958-57-4
Health Hazard 1H314
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c