Isododecane, mixture of isomers, tech. 80%
Isododecane, mixture of isomers, tech. 80%
Isododecane, mixture of isomers, tech. 80%
Isododecane, mixture of isomers, tech. 80%
Thermo Scientific Chemicals
Isododecane, mixture of isomers, tech. 80%
CAS: 31807-55-3 | C12H26 | 170.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2500 mL
Catalog number ALFA14235.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialIsododecane, Mixture of Isomers
Name Notemixture of isomers
CAS31807-55-3
Health Hazard 1H226-H304
Health Hazard 2GHS H Statement
H304-H226
View more
Isododecane, mixture of isomer acts as a reaction medium used for polymerization reactions. It is also useful as an emollient and solvent in skin care products due to its high spreadability, low viscosity and density. It is commonly used in anti-aging serums and also useful in many different cosmetic items like eyeliner, hair care, hair sprays, perfume, conditioners and lotions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isododecane, mixture of isomer acts as a reaction medium used for polymerization reactions. It is also useful as an emollient and solvent in skin care products due to its high spreadability, low viscosity and density. It is commonly used in anti-aging serums and also useful in many different cosmetic items like eyeliner, hair care, hair sprays, perfume, conditioners and lotions.
Solubility
Immiscible with water.
Notes
Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Chan, N. H.; Hizaddin, H. F.; Anantharaj, R.; Banerjee, T. Ethylsulphate-Based Ionic Liquids in the Liquid-Liquid Extraction of Pyrrole and Pyridine from Isododecane at 298.15 K. Chem. Prod. Process Model. 2015, 10 (3), 161-171.
  2. Ayesterán, J.; Kopriwa, N.; Buchbender, F.; Kalem, M.; Pfennig, A. ReDrop-A Simulation Tool for the Design of Extraction Columns Based on Single-Drop Experiments. Chem. Eng. Technol. 2015, 38 (10), 1894-1900.