Sebacic acid, 98+%
Sebacic acid, 98+%
Sebacic acid, 98+%
Thermo Scientific Chemicals
Sebacic acid, 98+%
CAS: 111-20-6 | C10H18O4 | 202.25 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14158.0B1000 g
Catalog number ALFA14158.0B
Price (MYR)
876.00
EA
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (MYR)
876.00
EA
Add to cart
Specifications
Chemical Name or MaterialSebacic acid
CAS111-20-6
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
Sebacic acid is used as a monomer for nylon 610, plasticizers, lubricants, hydraulic fluids, cosmetics and candles. It is also used as intermediates in antiseptics, aromatics and painting materials. It is used in the synthesis of poly(glycerol sebacate) and novel bio-nylon. It finds application as adhesives, corrosion inhibitors and solvents. Its derivative, diethyl Sebacate is used in plastics manufacturing and engineering, as well as drug delivery procedures through the use of biodegradable encapsulations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sebacic acid is used as a monomer for nylon 610, plasticizers, lubricants, hydraulic fluids, cosmetics and candles. It is also used as intermediates in antiseptics, aromatics and painting materials. It is used in the synthesis of poly(glycerol sebacate) and novel bio-nylon. It finds application as adhesives, corrosion inhibitors and solvents. Its derivative, diethyl Sebacate is used in plastics manufacturing and engineering, as well as drug delivery procedures through the use of biodegradable encapsulations.
Solubility
Slightly soluble in water.
Notes
Incompatible with bases, reducing agents and oxidizing agents.
RUO – Research Use Only
General References:
  1. Yabalak, E.; Gormez, O.; Gozmen, B.; Gizir, A. M. The solubility of sebacic acid in subcritical water using the response surface methodology. Int. J. Ind. Chem. 2015, 6 (1), 23-29.
  2. Kang, Y.; Chen, Z.; Wang, B.; Yang, Y. Synthesis and mechanical properties of thermoplastic films from lignin, sebacic acid and poly (ethylene glycol). Ind. Crops Prod. 2014, 56, 105-112.