1,10-Phenanthroline monohydrate, 99+%
1,10-Phenanthroline monohydrate, 99+%
1,10-Phenanthroline monohydrate, 99+%
1,10-Phenanthroline monohydrate, 99+%
Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, 99+%
CAS: 5144-89-8 | C12H10N2O | 198.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number ALFA14140.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material1,10-Phenanthroline monohydrate
CAS5144-89-8
Health Hazard 1H301
Health Hazard 2GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3P264b-P270-P301+P310-P330-P501c
View more
1,10-Phenanthroline monohydrate is useful to study the DNA-protein interactions due to its nuclease activity. It is used as a redox indicator and reagent for spectrophotometric assay of silver. It acts as a chelating ligand for the determination of Fe, Pd and V. It also acts as a matrix metalloproteinase inhibitor. It is a photometric reagent for iron, which forms a red chelate with ferrous (iron II) ion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,10-Phenanthroline monohydrate is useful to study the DNA-protein interactions due to its nuclease activity. It is used as a redox indicator and reagent for spectrophotometric assay of silver. It acts as a chelating ligand for the determination of Fe, Pd and V. It also acts as a matrix metalloproteinase inhibitor. It is a photometric reagent for iron, which forms a red chelate with ferrous (iron II) ion.
Solubility
Soluble in ethanol, acetone and methanol. Slightly soluble in water and benzene.
Notes
Working solutions of 1,10-Phenanthroline monohydrate may stable for a few days at refrigerated temperatures. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
General References:
  1. Redox indicator.
  2. Chelating ligand for the determination of Fe, Pd and V.
  3. Said, N.; Muhammad, R. S.; Qamar, A.; Zahid, H. One Pot Synthesis of 1,10-Phenanthroline-based Shape-Persistent Flouroscent Macrocycle Using Sonogashira Coupling. Lett. Org. Chem. 2015, 12 (7), 504-510.