1-Bromonaphthalene, 97%
1-Bromonaphthalene, 97%
1-Bromonaphthalene, 97%
1-Bromonaphthalene, 97%
Thermo Scientific Chemicals
1-Bromonaphthalene, 97%
CAS: 90-11-9 | C10H7Br | 207.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1000 g
Catalog number ALFA14122.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Bromonaphthalene
CAS90-11-9
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Health Hazard 3P264b-P270-P301+P312-P330-P501c
View more
1-Bromonaphthalene is used in organic synthesis and refractometry of fats. It is used in the preparation of N-aryl imidazoles and diaryl ethers. It is used to determine the refractive index of crystals and the water content of alcohols. It is utilized in the preparation of glutathione peroxidase-like antioxidant activity of diaryl diselenides. Further, it serves as a solvent for the exfoliation and dispersion of hexabenzocoronene. In addition to this, it is used as a refrigerant and a solvent molecular weight substance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromonaphthalene is used in organic synthesis and refractometry of fats. It is used in the preparation of N-aryl imidazoles and diaryl ethers. It is used to determine the refractive index of crystals and the water content of alcohols. It is utilized in the preparation of glutathione peroxidase-like antioxidant activity of diaryl diselenides. Further, it serves as a solvent for the exfoliation and dispersion of hexabenzocoronene. In addition to this, it is used as a refrigerant and a solvent molecular weight substance.
Solubility
Miscible with chloroform, ethanol, ether, benzene and ethyl acetate. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Motogaito, A.; Nakamura, S.; Miyazaki, J.; Miyake, H.; Hiramatsu, K. Using surface-plasmon polariton at the GaP-Au interface in order to detect chemical species in high-refractive-index media. Opt. Commun. 2015, 341, 64-68.
  2. Gentry, S. P.; Halloran, J. W. Light scattering in absorbing ceramic suspensions: Effect on the width and depth of photopolymerized features. J. Eur. Ceram. Soc. 2015, 35 (6), 1895-1904.