2,4-Difluorobenzonitrile, 98%
2,4-Difluorobenzonitrile, 98%
2,4-Difluorobenzonitrile, 98%
2,4-Difluorobenzonitrile, 98%
Thermo Scientific Chemicals
2,4-Difluorobenzonitrile, 98%
CAS: 3939-09-1 | C7H3F2N | 139.11 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14113.1425 g
Catalog number ALFA14113.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material2,4-Difluorobenzonitrile
CAS3939-09-1
Health Hazard 1H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c