2-Mercaptobenzothiazole, 97%
2-Mercaptobenzothiazole, 97%
2-Mercaptobenzothiazole, 97%
2-Mercaptobenzothiazole, 97%
Thermo Scientific Chemicals
2-Mercaptobenzothiazole, 97%
CAS: 149-30-4 | C7H5NS2 | 167.244 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA14086.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Mercaptobenzothiazole
CAS149-30-4
Health Hazard 1H317
Health Hazard 2GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.
Health Hazard 3P261-P272-P280g-P302+P352-P333+P313-P363-P501c
View more
2-Mercaptobenzothiazole is used for detection of the equivalence point in volumetry (complexometry) and for concentration determinations in colorimetry. It is also used for the photometric determination of gold, cadmium, copper, lead and palladium. Further, it is used as a metalworking fluid, antifreeze, grease, anticorrosive agent, and adhesive. It acts as an antibacterial and antifungal. It is also employed in the rubber industry as a vulcanization accelerator. It inhibits dopamine beta-hydroxylase and biocorrosion in cooling systems, especially in the paper industry. In addition to this, it is used as a wood preservative and rubber additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Mercaptobenzothiazole is used for detection of the equivalence point in volumetry (complexometry) and for concentration determinations in colorimetry. It is also used for the photometric determination of gold, cadmium, copper, lead and palladium. Further, it is used as a metalworking fluid, antifreeze, grease, anticorrosive agent, and adhesive. It acts as an antibacterial and antifungal. It is also employed in the rubber industry as a vulcanization accelerator. It inhibits dopamine beta-hydroxylase and biocorrosion in cooling systems, especially in the paper industry. In addition to this, it is used as a wood preservative and rubber additive.
Solubility
Soluble in ethanol, diethyl ether, benzene, carbon tetrachloride, alkali and alkaline solutions. Slightly soluble in acetone, naphtha and glacial acetic acid. Insoluble in water and gasoline.
Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Reagent for determination of Ag, Au, Bu, Cd, Hg, Ir, Pt, Tl.
  2. Gries, W.; Küpper, K.; Leng, G. Rapid and sensitive LC-MS-MS determination of 2-mercaptobenzothiazole, a rubber additive, in human urine. Anal. Bioanal. Chem. 2015, 407 (12), 3417-3423.
  3. Balaskas, A. C.; Curioni, M.; Thompson, G. E. Effectiveness of 2-mercaptobenzothiazole, 8-hydroxyquinoline and benzotriazole as corrosion inhibitors on AA 2024-T3 assessed by electrochemical methods. Surf. Interface Anal. 2015, 47 (11), 1029-1039.