4-Aminothiophenol, 97%
4-Aminothiophenol, 97%
4-Aminothiophenol, 97%
4-Aminothiophenol, 97%
Thermo Scientific Chemicals
4-Aminothiophenol, 97%
CAS: 1193-02-8 | C6H6NS | 124.18 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14082.065 g
Catalog number ALFA14082.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Aminothiophenol
CAS1193-02-8
Health Hazard 1H302-H314
Health Hazard 2GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
View more
4-Aminothiophenol is used in the synthesis of pectin-4-aminothiophenol conjugates and it forms self-assembled monolayers (SAMs) in the development of DNA. It is used in the detection of thrombin by protein recognition using surface-enhanced Raman spectroscopy (SERS), which finds applications in silver and gold nano particles. It acts as a linker to thiol (SH) functionalizes multiwalled carbon nanotubes (MWCNTs).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Aminothiophenol is used in the synthesis of pectin-4-aminothiophenol conjugates and it forms self-assembled monolayers (SAMs) in the development of DNA. It is used in the detection of thrombin by protein recognition using surface-enhanced Raman spectroscopy (SERS), which finds applications in silver and gold nano particles. It acts as a linker to thiol (SH) functionalizes multiwalled carbon nanotubes (MWCNTs).
Solubility
Immiscible with water.
Notes
Air sensitive. Store in a cool place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Cao, B.; Liu, B.; Yang, J. Facile synthesis of single crystalline gold nanoplates and SERS investigations of 4-aminothiophenol. Cryst. Eng. Comm. 2013, 15, 5735-5738.
  2. Yue, G.; Li, F.; Tan, F.; Li, G.; Chen, C.; Wu, J. Nickel sulfide films with significantly enhanced electrochemical performance induced by self-assembly of 4-aminothiophenol and their application in dye-sensitized solar cells. RSC Adv. 2014, 4, 64068-64074.