5-Chlorosalicylaldehyde, 98%
5-Chlorosalicylaldehyde, 98%
5-Chlorosalicylaldehyde, 98%
5-Chlorosalicylaldehyde, 98%
Thermo Scientific Chemicals
5-Chlorosalicylaldehyde, 98%
CAS: 635-93-8 | C7H5ClO2 | 156.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFA14069.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material5-Chlorosalicylaldehyde
CAS635-93-8
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c