Benzoic acid, 99%
Benzoic acid, 99%
Benzoic acid, 99%
Thermo Scientific Chemicals
Benzoic acid, 99%
CAS: 65-85-0 | C7H6O2 | 122.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFA14062.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBenzoic Acid
CAS65-85-0
Melting Point121°C to 124°C
Boiling Point249°C
Health Hazard 1H315-H318-H372-H500
View more
Benzoic acid is an important precursor for the preparation of many other organic substances viz. benzoyl chloride, benylbenzoate, phenol, sodium benzoate, benzyl alcohol as well as benzoate plasticizers such as glycol-, diethhyleneglycol- and triethyleneglycol esters. Benzoic acid and its salts are used as food preservatives. It is involved in rubber polymerization as an activator and a retardant. It is the main component of benzoin resin and is a constituent of Whitfield’s ointment, which is used for the treatment of fungal skin diseases such as tinea, ringworm and athlete’s foot. It is widely used in cosmetics, dyes, plastics and insect repellents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in acetone, benzene, chloroform, alcohol, diethyl ether, hexane and liquid ammonia. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents, reducing agents and strong bases.
RUO – Research Use Only
General References:
  1. Sarcevica, I.; Orola, L.; Nartowski, K. P.; Khimyak, Y. Z.; Round, A. N.; Fabian, L. Mechanistic and Kinetic Insight into Spontaneous Cocrystallization of Isoniazid and Benzoic Acid. Mol. Pharmaceutics 2015, 12 (8), 2981-2992.
  2. Osowole, A. A.; Wakil, S. M. Synthesis, characterization, antioxidant and antimicrobial activities of someMetal(II) Complexes of the Mixed-Ligands, Vitamin B2 and Benzoic acid. Appl. Chem. 2015, 79, 30370-30374.