1-Bromo-3,5-difluorobenzene, 98%
1-Bromo-3,5-difluorobenzene, 98%
1-Bromo-3,5-difluorobenzene, 98%
1-Bromo-3,5-difluorobenzene, 98%
Thermo Scientific Chemicals
1-Bromo-3,5-difluorobenzene, 98%
CAS: 461-96-1 | C6H3BrF2 | 192.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
10 g
25 g
Catalog number ALFA14053.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Bromo-3,5-difluorobenzene
CAS461-96-1
Health Hazard 1H226-H302-H315-H317-H319-H373
Health Hazard 2GHS H Statement
H226-H373-H302-H315-H317
Flammable liquid and vapour.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Health Hazard 3P210-P233-P240-P241-P242-P243-P260-P264b-P270-P272-P280-P301+P312-P303+P361+P353-P305+P351+P338-P314-P330-P333+P313-P363-P370+P378q-P501c