Sodium phenylpyruvate monohydrate, 98%
Sodium phenylpyruvate monohydrate, 98%
Sodium phenylpyruvate monohydrate, 98%
Sodium phenylpyruvate monohydrate, 98%
Thermo Scientific Chemicals
Sodium phenylpyruvate monohydrate, 98%
CAS: 122049-54-1 | C9H7O3 | 163.15 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14027.031 g
Catalog number ALFA14027.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium phenylpyruvate monohydrate
CAS122049-54-1
Melting Point>300°C
Recommended StorageKeep cold
EINECS Number204-053-2
View more
Sodium phenylpyruvate monohydrate is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium phenylpyruvate monohydrate is used as an organic chemical synthesis intermediate.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Zhiwei Guo; Animesh Goswami; K.David Mirfakhrae; Ramesh N.Patel. Asymmetric acyloin condensation catalyzed by phenylpyruvate decarboxylase. Tetrahedron: Asymmetry. 1999, 10,(24), 4667-4675.
  2. Jeffrey D.Hermes; Paul M.Weiss; W.W.Cleland. Use of nitrogen-15 and deuterium isotope effects to determine the chemical mechanism of phenylalanine ammonia-lyase. Biochemistry. 1985, 24,(12), 2959-2967.