Glycerol monoacetate, tech., mixture of isomers, cont. varying amounts of diacetate
Glycerol monoacetate, tech., mixture of isomers, cont. varying amounts of diacetate
Glycerol monoacetate, tech., mixture of isomers, cont. varying amounts of diacetate
Glycerol monoacetate, tech., mixture of isomers, cont. varying amounts of diacetate
Thermo Scientific Chemicals
Glycerol monoacetate, tech., mixture of isomers, cont. varying amounts of diacetate
CAS: 26446-35-5 | C5H10O4 | 134.13 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14025.30250 g
Catalog number ALFA14025.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGlycerol monoacetate, tech.
Name Notemixture of isomers
CAS26446-35-5
Recommended StorageAmbient temperatures
Density1.21
View more
Glycerol monoacetate is used for production of tanning leather and dye. It is also used as a solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glycerol monoacetate is used for production of tanning leather and dye. It is also used as a solvent.
Solubility
Soluble in water, methanol, ethanol, ether (slightly), and insoluble in benzene.
Notes
Hygroscopic. Protect from moisture. Store at room temperature. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. R.A. Peters; P.L. Morselli. Observations on the use and action of monoacetin in fluoroacetate poisoning. Biochemical Pharmacology. 1965, 14 (12), 1891-1893.
  2. Takuya Fukumura; Takuji Toda; Yuichiro Seki; Masaki Kubo; Naomi Shibasaki-Kitakawa and Toshikuni Yonemoto. Catalytic Synthesis of Glycerol Monoacetate Using a Continuous Expanded Bed Column Reactor Packed with Cation-Exchange Resin. Ind. Eng. Chem. Res. 2009, 48 (4), 1816-1823.