4-Isopropylbenzaldehyde, tech. 90%
4-Isopropylbenzaldehyde, tech. 90%
4-Isopropylbenzaldehyde, tech. 90%
4-Isopropylbenzaldehyde, tech. 90%
Thermo Scientific Chemicals
4-Isopropylbenzaldehyde, tech. 90%
CAS: 122-03-2 | C10H12O | 148.21 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14019.22100 g
Catalog number ALFA14019.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Isopropylbenzaldehyde
CAS122-03-2
Health Hazard 1H227-H302
Health Hazard 2GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c