1,4-Diazabicyclo[2.2.2]octane, 98%
1,4-Diazabicyclo[2.2.2]octane, 98%
1,4-Diazabicyclo[2.2.2]octane, 98%
1,4-Diazabicyclo[2.2.2]octane, 98%
Thermo Scientific Chemicals
1,4-Diazabicyclo[2.2.2]octane, 98%
CAS: 280-57-9 | C6H12N2 | 112.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA14003.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material1,4-Diazabicyclo[2.2.2]octane
CAS280-57-9
Health Hazard 1H228-H302-H315-H318-H500
Health Hazard 2GHS H Statement
H228-H318-H302-H335-H315
Flammable solid.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3P210-P240-P241-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P332+P313-P362-P370+P378q-P501c
View more
1,4-Diazabicyclo[2.2.2]octane is used as polyurethane catalyst, Balis-Hillman reaction catalyst complexing ligand and lewis base. It finds use in dye lasers and in mounting samples for fluorescence microscopy and as anti-fade reagent shown to scavenge free radicals due to flurochrome excitation of fluorochromes. Further, it is an oxidation and polymerization catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,4-Diazabicyclo[2.2.2]octane is used as polyurethane catalyst, Balis-Hillman reaction catalyst complexing ligand and lewis base. It finds use in dye lasers and in mounting samples for fluorescence microscopy and as anti-fade reagent shown to scavenge free radicals due to flurochrome excitation of fluorochromes. Further, it is an oxidation and polymerization catalyst.
Solubility
Soluble in water, acetone, benzene, ethanol and methyl ethyl ketone.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
General References:
  1. Dabco is a registered trademark of Air Products and Chemicals Inc.
  2. Forms crystalline complexes with organolithium compounds: J. Am. Chem. Soc., 87, 3276 (1965), which show enhanced reactivity, e.g. in the high yield ɑ-metallation of thioanisole by n-BuLi: J. Org. Chem., 31, 4097 (1966).
  3. Forms a stable crystalline complex with H2O2, useful as an equivalent to anhydrous H2O2: reaction with TMS chloride gives bis(TMS) peroxide, a useful OH+-equivalent, e.g. in the conversion of aryllithiums to phenols: Synthesis, 633 (1986).