Disodium hydrogen citrate sesquihydrate, 99%
Disodium hydrogen citrate sesquihydrate, 99%
Disodium hydrogen citrate sesquihydrate, 99%
Disodium hydrogen citrate sesquihydrate, 99%
Thermo Scientific Chemicals
Disodium hydrogen citrate sesquihydrate, 99%
CAS: 6132-05-4 | C12H18Na4O17 | 526.22 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14001.30250 g
Catalog number ALFA14001.30
Price (MYR)
312.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
312.00
EA
Add to cart
Specifications
Chemical Name or MaterialDisodium hydrogen citrate sesquihydrate
CAS6132-05-4
Melting Point149°C (decomposition)
Recommended StorageAmbient temperatures
EINECS Number205-623-3
View more
Disodium hydrogen citrate sesquihydrate is a reagent employed in the investigation of mediatorless laccase cathodes as half-cell electrode under galvanostatic load by using a two-compartment set up. It is used in the extraction of organ/tissue sample.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Disodium hydrogen citrate sesquihydrate is a reagent employed in the investigation of mediatorless laccase cathodes as half-cell electrode under galvanostatic load by using a two-compartment set up. It is used in the extraction of organ/tissue sample.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Huertas-Pérez, J. F.; Arroyo-Manzanares, N.; García-Campaña, A. M.; Gámiz-Gracia, L. High-throughput determination of citrinin in rice by ultra-high-performance liquid chromatography and fluorescence detection (UHPLC-FL). Food Addit. Contam. Part A 2015, 32 (8), 1352-1357.
  2. Masiá, A.; Vásquez, K.; Campo, J.; Picó, Y. Assessment of two extraction methods to determine pesticides in soils, sediments and sludges. Application to the Túria River Basin. J. Chromatogr. A 2015, 1378, 19-31.