Perfluorononanoic acid, 97%
Perfluorononanoic acid, 97%
Perfluorononanoic acid, 97%
Perfluorononanoic acid, 97%
Perfluorononanoic acid, 97%
CAS: 375-95-1 | C9HF17O2 | 464.078 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13980.065 g
Catalog number ALFA13980.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPerfluorononanoic acid
CAS375-95-1
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.