2-Amino-4-nitrophenol, 98%, Thermo Scientific Chemicals
2-Amino-4-nitrophenol, 98%, Thermo Scientific Chemicals
2-Amino-4-nitrophenol, 98%, Thermo Scientific Chemicals
2-Amino-4-nitrophenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Amino-4-nitrophenol, 98%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA13964.1425 g
Catalog number ALFA13964.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Amino-4-nitrophenol
CAS99-57-0
Health Hazard 1H302-H312-H315-H317-H319-H332-H335-H340
Health Hazard 2GHS H Statement
H302-H319-H317
Harmful if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P261-P264b-P270-P271-P272-P280-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P501c