N,N'-Bis(salicylidene)ethylenediamine, 98%
N,N'-Bis(salicylidene)ethylenediamine, 98%
N,N'-Bis(salicylidene)ethylenediamine, 98%
N,N'-Bis(salicylidene)ethylenediamine, 98%
Thermo Scientific Chemicals
N,N'-Bis(salicylidene)ethylenediamine, 98%
CAS: 94-93-9 | C16H16N2O2 | 268.32 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13885.1425 g
Catalog number ALFA13885.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN,N'-Bis(salicylidene)ethylenediamine
CAS94-93-9
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c