1,2-Dichlorobenzene, 99%
1,2-Dichlorobenzene, 99%
1,2-Dichlorobenzene, 99%
1,2-Dichlorobenzene, 99%
Thermo Scientific Chemicals
1,2-Dichlorobenzene, 99%
CAS: 95-50-1 | C6H4Cl2 | 146.998 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13881.0E2500 g
Catalog number ALFA13881.0E
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2500 g
Request bulk or custom format
Specifications
Chemical Name or Material1,2-Dichlorobenzene
CAS95-50-1
Health Hazard 1H227-H302+H332-H315-H317-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P210-P235-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P370+P378q-P501c
View more
1,2-Dichlorobenzene is widely used as a precursor to prepare 1,2-dichloro-4-nitrobenzene, which is an intermediate in the synthesis of agrochemicals. It acts as a solvent for fullerenes. It is also used in softening and removing carbon-based contamination on metal surfaces. It also used as food preservative, motor oil additive, lubricant and paint.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2-Dichlorobenzene is widely used as a precursor to prepare 1,2-dichloro-4-nitrobenzene, which is an intermediate in the synthesis of agrochemicals. It acts as a solvent for fullerenes. It is also used in softening and removing carbon-based contamination on metal surfaces. It also used as food preservative, motor oil additive, lubricant and paint.
Solubility
Miscible with water and most of the organic solvents.
Notes
Keep away from sources of ignition. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Chen, N. Y.; Yang, S. C.; Liu, M. C.; Lee, J. F.; Chang, J. R. Pellet Vanadia Catalysts for Oxidative Destruction of 1,2-Dichlorobenzene: Roles of the Grafted TiO2 in Vanadia Morphology and Catalytic Reaction. Catal. Surv. Asia 2015, 19 (1), 38-56.
  2. Cai, T.; Huang, H.; Deng, W.; Dai, Q.; Liu, W.; Wang, X. Catalytic combustion of 1, 2-dichlorobenzene at low temperature over Mn-modified Co3O4 catalysts. Appl. Catal., B 2015, 166-167, 393-405.