3-Acetyl-2,5-dichlorothiophene, 98%
3-Acetyl-2,5-dichlorothiophene, 98%
3-Acetyl-2,5-dichlorothiophene, 98%
3-Acetyl-2,5-dichlorothiophene, 98%
Thermo Scientific Chemicals
3-Acetyl-2,5-dichlorothiophene, 98%
CAS: 36157-40-1 | C6H4Cl2OS | 195.057 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13867.065 g
Catalog number ALFA13867.06
Price (MYR)
496.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
496.00
EA
Add to cart
Specifications
Chemical Name or Material3-Acetyl-2,5-dichlorothiophene
CAS36157-40-1
Health Hazard 1H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c