2-Bromo-4-methylaniline, 99%
2-Bromo-4-methylaniline, 99%
2-Bromo-4-methylaniline, 99%
2-Bromo-4-methylaniline, 99%
Thermo Scientific Chemicals
2-Bromo-4-methylaniline, 99%
CAS: 583-68-6 | C7H8BrN | 186.052 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13818.0910 g
Catalog number ALFA13818.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Bromo-4-methylaniline
CAS583-68-6
Health Hazard 1H301+H311+H331-H315-H319-H335-H373
Health Hazard 2GHS H Statement
H301-H311-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P314-P330-P332+P313-P361-P363-P501c
View more
The N-(3, 5-Dichlorophenyl) naphthaldimine was obtained from the reaction of 2-hy- droxynaphthalene-l-carbaldehyde (0.01 mol) with a solution of 2-bromo-4-methylaniline (0.01 mol) in 40 ml of ethanol. Reaction of 2-bromo-4-methylaniline was quite clean, albeit 3,4-diaminotoluene was isolated just in 37% yield after flash chromatography on silica gel. The bromomethylquinoline starting material 16 was synthesized in two steps using an optimized process route by condensing 2-bromo-4-methylaniline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The N-(3, 5-Dichlorophenyl) naphthaldimine was obtained from the reaction of 2-hy- droxynaphthalene-l-carbaldehyde (0.01 mol) with a solution of 2-bromo-4-methylaniline (0.01 mol) in 40 ml of ethanol. Reaction of 2-bromo-4-methylaniline was quite clean, albeit 3,4-diaminotoluene was isolated just in 37% yield after flash chromatography on silica gel. The bromomethylquinoline starting material 16 was synthesized in two steps using an optimized process route by condensing 2-bromo-4-methylaniline.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Renee Aspiotis,; Denis Deschênes,; Daniel Dubé,; Yves Girard,; Zheng Huang,; France Laliberté,; Susana Liu,; Robert Papp,; Donald W. Nicholson,; Robert N. Young.The discovery and synthesis of highly potent subtype selective phosphodiesterase 4D inhibitors. Bioorganic & Medicinal Chemistry Letters 2010, 20(18), 5502-5505.
  2. A. Elmali,; Y. Elerman,; I. Svoboda and H. Fuess. N-(3,5-Dichlorophenyl)naphthaldimine. Acta Cryst.. 1998, c54 974-976.