Ammonium molybdate (para) tetrahydrate, 99%
Ammonium molybdate (para) tetrahydrate, 99%
Ammonium molybdate (para) tetrahydrate, 99%
Ammonium molybdate (para) tetrahydrate, 99%
Thermo Scientific Chemicals
Ammonium molybdate (para) tetrahydrate, 99%
CAS: 12054-85-2 | (NH4)6Mo7O24·4H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1000 g
Catalog number ALFA13766.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium molybdate (para) tetrahydrate
Melting Point∼190°C (decomposition)
CAS12054-85-2
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Ammonium molybdate tetrahydrate is widely utilized in cryo-negative staining which helps to visualize particles while preserving the sample. It is used as a catalyst to desulfurize thiophene and its derivatives. It serves as an analytical reagent to measure the amount of phosphates, silicates, arsenates and lead in an aqueous solution. It is also used in electroplating, in fertilizers for crops and in the production of molybdenum metal and ceramics. It is used as an analgesic in medical field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ammonium molybdate tetrahydrate is widely utilized in cryo-negative staining which helps to visualize particles while preserving the sample. It is used as a catalyst to desulfurize thiophene and its derivatives. It serves as an analytical reagent to measure the amount of phosphates, silicates, arsenates and lead in an aqueous solution. It is also used in electroplating, in fertilizers for crops and in the production of molybdenum metal and ceramics. It is used as an analgesic in medical field.
Solubility
Soluble in water. Insoluble in alcohol.
Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
General References:
  1. The combination with hydrogen peroxide is a system for the ambient temperature oxidative cleavage of epoxides to form ɑ-hydroxy ketones: Chem. Lett., 844 (2000).
  2. Lince, J. R.; Pluntze, A. M.; Jackson, A.; Radhakrishnan, G.; Adams, P. M. Tribochemistry of MoS3Nanoparticle Coatings. Tribol Lett. 2014, 53 (3), 543-554.
  3. McIntosha, S.; Bosscheb, M. V. Towards Understanding the Hydrocarbon Oxidation Activity of Oxides for Direct Hydrocarbon SOFC Anodes. ECS Trans. 2011, 35 (1), 2781-2788.