Geraniol, 97%
Geraniol, 97%
Geraniol, 97%
Thermo Scientific Chemicals
Geraniol, 97%
CAS: 106-24-1 | C10H18O | 154.253 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALFA13736.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGeraniol
CAS106-24-1
Health Hazard 1H227-H315-H317-H318
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P210-P261-P264b-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P362-P370+P378q-P501c
View more
Geraniol is used in the synthesis of insect repellant. It is also used in the synthesis of Angelicoin A and Herecinone J, which inhibit collagen-induced platelet aggregation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Geraniol is used in the synthesis of insect repellant. It is also used in the synthesis of Angelicoin A and Herecinone J, which inhibit collagen-induced platelet aggregation.
Solubility
Not miscible in water.
Notes
Incompatible with Strong oxidizing agents, acid chlorides, acid anhydrides. Keep away from heat. Air sensitive.
RUO – Research Use Only
General References:
  1. J. M. kim; M. R. Marshall; J.A. Cornell; J. F. Preston and C. I. wei. Antibacterial Activity of Carvacrol, Citral, and Geraniol against Salmonella typhimurium in Culture Medium and on Fish Cubes.Journal of Food Science.1995, 60 1364-1368.
  2. Yvette D. Burke; M. Jennifer Stark; Steven L. Roach; Stephanie E. Sen; Pamela L. Crowell. Inhibition of pancreatic cancer growth by the dietary isoprenoids farnesol and geraniol. Lipids.1997, 32 151-156.