L-Isoleucine, 99%
L-Isoleucine, 99%
L-Isoleucine, 99%
Thermo Scientific Chemicals
L-Isoleucine, 99%
L-isoleucine, CAS # 73-32-5, is a branched-chain aliphatic amino acid that is an essential constituent of the diet and is also used in several biological mechanisms, including the biosynthesis of proteins.
Have Questions?
Catalog NumberQuantity
ALFA13699.1850 g
Catalog number ALFA13699.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL-Isoleucine
Melting Point∼287°C (decomposition)
CAS73-32-5
Recommended StorageAmbient temperatures
RTECS NumberNR4705000
View more
L-Isoleucine acts as an essential amino acid in human. It is used as a flavoring agent and nutritional supplement for foods. It is also used as an antipsychotic. It is employed as a component of proteins. Further, it stimulates immune function and promotes secretion of several hormones. In addition to this, it is used for hemoglobin formation and regulating blood sugar and energy levels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• L-isoleucine is an essential amino acid that organisms are incapable of synthesizing. It must be ingested in the diet and can be found mainly in meats, fish, cheese, eggs, seeds, and nuts
• L-Isoleucine is an isomer of L-leucine. It is synthesized from threonine and is a branched-chain hydrophobic amino acid
Application
• L-isoleucine is used in several biological mechanisms, including assisting wound healing, detoxifying nitrogenous wastes, stimulating immune function, and promoting hormone secretion
• L-isoleucine shows antipsychotic activity
• It is important for hemoglobin formation and regulating blood sugar and energy levels
• L-isoleucine participates in protein synthesis by binding with transfer RNA (tRNA) in the cell cytoplasm
• This compound also plays an important role as a metabolite for several species, including Saccharomyces cerevisiae, Escherichia coli, plants, algae, and mice
• L-isoleucine can be used as an ingredient for the development of other essential biochemical components in the organism, some of which are used for energy production and stimulants in the upper brain
• It has been used as a supplement of maintenance media (DMEM/F-12) for the culture of primary hepatocytes
• It has been used in capillary feeder assay to determine the consumption of amino acids by Drosophila melanogaster
RUO – Research Use Only
General References:
  1. Han, G.; Hu, X.; Wang, X. Co-production of S-adenosyl-L-methionine and L-isoleucine in Corynebacterium glutamicum. Enzyme Microb. Technol. 2015, 78, 27-33.
  2. Yin, L.; Zhao, J.; Chen, C.; Hu, X.; Wang, X. Enhancing the carbon flux and NADPH supply to increase L-isoleucine production in Corynebacterium glutamicum. Biotechnol. Bioprocess Eng. 2014, 19 (1), 132-142.
  3. Uchizono, s.; Tabuki, Y.; Kawaguchi, N.; Tanimura, T.; Itoh, T. Mated Drosophila melanogaster females consume more amino acids during the dark phase. PLoS One. 2017, 12, (2), e0172886.
  4. Plata-Gómez, A.; Crespo,M.; Arregui, C. Protocol for the assessment of mTOR activity in mouse primary hepatocytes. STAR Protoc. 2021, 2, (4), 100918.