Chlorotrimethylsilane, 98%
Chlorotrimethylsilane, 98%
Chlorotrimethylsilane, 98%
Chlorotrimethylsilane, 98%
Thermo Scientific Chemicals
Chlorotrimethylsilane, 98%
CAS: 75-77-4 | C3H9ClSi | 108.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
100 mL
Catalog number ALFA13651.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Boiling Point56°C to 58°C
Chemical Name or MaterialChlorotrimethylsilane
Melting Point-58°C
CAS75-77-4
Health Hazard 1H225-H301-H312+H332-H314
View more
Chlorotrimethylsilane is used to silanize laboratory glassware, which makes the surfaces more lipophilic. It is used in the production of trimethylsilyl halides, pseudohalides and various organic silicon compounds. It is also used to produce hexamethyldisilane by reduction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chlorotrimethylsilane is used to silanize laboratory glassware, which makes the surfaces more lipophilic. It is used in the production of trimethylsilyl halides, pseudohalides and various organic silicon compounds. It is also used to produce hexamethyldisilane by reduction.
Solubility
Miscible with ether, benzene, diethylether and perchloroethylene.
Notes
Incompatible with water, strong acids, strong oxidizing agents, strong bases, alcohols, aldehydes, amines, ketones and esters.
RUO – Research Use Only
General References:
  1. Feng, S.; Gu, Z.; Tang, Y.; Liu, H.; Jiang, G. An efficient and green synthetic route to losartan. J. Chem. Res. 2015, 39 (8), 433-494.
  2. Horak, T.; Sterba, K.; Olsovska, J. Quantitative determination of fluorine in spent grain and brewery yeast. J. Inst. Brew. 2015, 121 (2), 193-196.