3-Dimethylaminobenzoic acid, 98%
3-Dimethylaminobenzoic acid, 98%
3-Dimethylaminobenzoic acid, 98%
3-Dimethylaminobenzoic acid, 98%
3-Dimethylaminobenzoic acid, 98%
CAS: 99-64-9 | C9H11NO2 | 165.192 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13612.22100 g
Catalog number ALFA13612.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Dimethylaminobenzoic acid
CAS99-64-9
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Melting Point148°C to 153°C
View more
3-Dimethylaminobenzoic acid is used as a benzoic acid derivative.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Dimethylaminobenzoic acid is used as a benzoic acid derivative.
Solubility
Soluble in methanol (50 mg/ml). Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to moisture. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. W.L. Baker.; Kunamalar Sabapathy.; Maria Vibat.; G. Lonergan. Laccase catalyzes formation of an indamine dye between 3-methyl-2-benzothiazolinone hydrazone and 3-dimethylaminobenzoic acid. Enzyme and Microbial Technology. 1996, 18 (2), 90-94.
  2. Padmarajaiah Nagaraja.; Anantharaman Shivakumar.; Ashwinee Kumar Shrestha. Peroxidase-Catalyzed Oxidative Coupling of Paraphenylenediamine with 3-Dimethylaminobenzoic Acid: Application in Crude Plant Extracts. J. Agric. Food Chem. 2007, 57 (12), 5173-5177.