3-Fluorophenol, 98%
3-Fluorophenol, 98%
3-Fluorophenol, 98%
Thermo Scientific Chemicals
3-Fluorophenol, 98%
CAS: 372-20-3 | C6H5FO | 112.103 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13598.0910 g
Catalog number ALFA13598.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Fluorophenol
CAS372-20-3
Health Hazard 1H227-H301-H312-H315-H318-H332-H335
Health Hazard 2GHS H Statement
H314-H318-H302-H312-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
Health Hazard 3P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c