4-Hydroxybenzaldehyde, 98%
4-Hydroxybenzaldehyde, 98%
4-Hydroxybenzaldehyde, 98%
4-Hydroxybenzaldehyde, 98%
Thermo Scientific Chemicals
4-Hydroxybenzaldehyde, 98%
CAS: 123-08-0 | C7H6O2 | 122.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ALFA13580.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Hydroxybenzaldehyde
CAS123-08-0
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
View more
4-Hydroxybenzaldehyde is used as a chemical that maintains bactericidal activity when tested against certain bacteria strains.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Hydroxybenzaldehyde is used as a chemical that maintains bactericidal activity when tested against certain bacteria strains.
Solubility
Soluble in water(13.8g/L), ether, alcohol, and acetone.
Notes
Air Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only
General References:
  1. Sarhan, A.A.; Abdelaal, M.Y.; Ali, M.M.; Abdel-Latiff, E.H. Reaction of crosslinked chloromethylated polystyrene with 4-hydroxybenzaldehyde under phase transfer-catalyzed conditions. Polymer. 1999, 40, (1), 233-241.
  2. Olga d'Hennezel.; Pierre Pichat.; David F Ollis. Benzene and toluene gas-phase photocatalytic degradation over H2O and HCL pretreated TiO2: by-products and mechanisms. Journal of Photochemistry and Photobiology A: Chemistry. 1998, 118, (3), 197-204.