(+)-Rutin trihydrate, 95%
(+)-Rutin trihydrate, 95%
(+)-Rutin trihydrate, 95%
(+)-Rutin trihydrate, 95%
Thermo Scientific Chemicals
(+)-Rutin trihydrate, 95%
CAS: 250249-75-3 | C27H36O19 | 664.566 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA13570.22
Price (MYR)
772.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
772.00
EA
Add to cart
Specifications
Chemical Name or Material(+)-Rutin trihydrate
CAS250249-75-3
Melting Point184°C (decomposition)
Recommended StorageAmbient temperatures
RTECS NumberVM2975000
View more
Rutin is the major mutagenic component of red wine, It is used in the activation of a mutagen by treatment both liver microsomal enzymes and fecal enzymes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Rutin is the major mutagenic component of red wine, It is used in the activation of a mutagen by treatment both liver microsomal enzymes and fecal enzymes.
Solubility
Soluble in DMSO, acetone, dichloromethane or ethyl acetate. Insoluble in water.
Notes
Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. William S. Schlotzhauer.; Robert M. Martin.; Maurice E. Snook.; Ralph E. Williamson. Pyrolytic studies on the contribution of tobacco leaf constituents to the formation of smoke catechols. J. Agric. Food Chem. 1982, 30, (2), 372-374.
  2. Jing Li.; Howard A. Chase. Characterization and evaluation of a macroporous adsorbent for possible use in the expanded bed adsorption of flavonoids from Ginkgo biloba L. Journal of Chromatography A. 2009, 1216, (50), 8730-8740.