n-Heptyl 4-hydroxybenzoate, 98%
n-Heptyl 4-hydroxybenzoate, 98%
n-Heptyl 4-hydroxybenzoate, 98%
n-Heptyl 4-hydroxybenzoate, 98%
Thermo Scientific Chemicals
n-Heptyl 4-hydroxybenzoate, 98%
CAS: 1085-12-7 | C14H20O3 | 236.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA13555.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Materialn-Heptyl 4-hydroxybenzoate
CAS1085-12-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
n-Heptyl 4-hydroxybenzoate is used for inhibiting microbiological spoilage, used as preservatives, skin care, hair care.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
n-Heptyl 4-hydroxybenzoate is used for inhibiting microbiological spoilage, used as preservatives, skin care, hair care.
Solubility
Soluble in alcohol, phenoxyethanol, propylene glycol. Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only