(1-Hexadecyl)pyridinium chloride monohydrate, 98%
(1-Hexadecyl)pyridinium chloride monohydrate, 98%
(1-Hexadecyl)pyridinium chloride monohydrate, 98%
(1-Hexadecyl)pyridinium chloride monohydrate, 98%
Thermo Scientific Chemicals
(1-Hexadecyl)pyridinium chloride monohydrate, 98%
CAS: 6004-24-6 | C21H40ClNO | 358.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA13499.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material(1-Hexadecyl)pyridinium chloride monohydrate
CAS6004-24-6
Health Hazard 1H301-H315-H318-H330-H335
Health Hazard 2GHS H Statement
H300-H330-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
View more
(1-Hexadecyl)pyridinium chloride monohydrate is used in a study to assess removal of emulsified oil from oily waste water and removal of emulsified food and mineral oils from waste water. And it is used for detergent-based extraction of membrane-associated enzymes such as alkaline phosphatase, and in extraction of chromium and palladium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(1-Hexadecyl)pyridinium chloride monohydrate is used in a study to assess removal of emulsified oil from oily waste water and removal of emulsified food and mineral oils from waste water. And it is used for detergent-based extraction of membrane-associated enzymes such as alkaline phosphatase, and in extraction of chromium and palladium.
Solubility
Soluble in water, alcohol, chloroform
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Hayati Filik.; Esma Tutem.; Resat Apak.; Erol Ercag. Spectrophotometric determination of gallium(III) with carminic acid and hexadecylpyridinium chloride. Microchimica Acta. 1998, 129, (1), 57-63.
  2. Renata Komendova-Vlasankova.; Lumír Sommer. Separation and Preconcentration of Platinum Group Metals and Gold on Modified Silica and XAD Sorbents in the Presence of Cationic Surfactants for Their Determination by ICP-AES. Collect. Czech. Chem. Commun. 2002, 67, (4), 454-470.
  3. Catalyst, used in combination with tungstophosphoric acid for the chemoselective oxidation of anilines to nitroso- or nitrobenzenes by hydrogen peroxide: J. Org. Chem., 58, 3633 (1993).