2,4-Difluorobenzoic acid, 98%
2,4-Difluorobenzoic acid, 98%
2,4-Difluorobenzoic acid, 98%
2,4-Difluorobenzoic acid, 98%
Thermo Scientific Chemicals
2,4-Difluorobenzoic acid, 98%
CAS: 1583-58-0 | C7H4F2O2 | 158.10 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13403.065 g
Catalog number ALFA13403.06
Price (MYR)
275.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
275.00
EA
Add to cart
Specifications
Chemical Name or Material2,4-Difluorobenzoic acid
CAS1583-58-0
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c