1-Methyl-4-piperidone, 98%
1-Methyl-4-piperidone, 98%
1-Methyl-4-piperidone, 98%
1-Methyl-4-piperidone, 98%
Thermo Scientific Chemicals
1-Methyl-4-piperidone, 98%
CAS: 1445-73-4 | C6H11NO | 113.16 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13352.1850 g
Catalog number ALFA13352.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Methyl-4-piperidone
CAS1445-73-4
Health Hazard 1H226-H314-H335
Health Hazard 2GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
View more
1-Methyl-4-piperidone, are widely used in artificial fibre industry. They are polymerizable and used as nylon precursors. They have excellent applications as solvents and as intermediates for organic synthesis. They are used in the aqueous carrier medium and amide penetrants in inks and water soluble paints.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Methyl-4-piperidone, are widely used in artificial fibre industry. They are polymerizable and used as nylon precursors. They have excellent applications as solvents and as intermediates for organic synthesis. They are used in the aqueous carrier medium and amide penetrants in inks and water soluble paints.
Solubility
It is miscible with water
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents, strong acids.
RUO – Research Use Only
General References:
  1. V Krishnakumar.; G Keresztury.; T Sundius. Density functional theory study of vibrational spectra and assignment of fundamental vibrational modes of 1-methyl-4-piperidone. Spectrochimica Acta Part . 2007, 68,(3), 845-850.
  2. Z Jia.; JW Quail.; VK Arora.; JR Dimmock. Structure of 3, 5-bis (benzylidene)-1-methyl-4-piperidone methobromide hemiethanol solvate. Acta Cryst. 1989, 45, 1117-1118 .