1,4-Diethylbenzene, 98%
1,4-Diethylbenzene, 98%
1,4-Diethylbenzene, 98%
1,4-Diethylbenzene, 98%
Thermo Scientific Chemicals
1,4-Diethylbenzene, 98%
CAS: 105-05-5 | C10H14 | 134.22 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13343.22100 g
Catalog number ALFA13343.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material1,4-Diethylbenzene
CAS105-05-5
Health Hazard 1H226-H304-H315-H318
Health Hazard 2GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3P210-P233-P240-P241-P242-P243-P264b-P280-P301+P310-P302+P352-P305+P351+P338-P310-P331-P332+P313-P362-P374-P380-P501c
View more
1,4-Diethylbenzene, is used as a solvent and an intermediate in the manufacture of other organic compounds for the finished products of such as curing agents, engineering plastics andcross-linking agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,4-Diethylbenzene, is used as a solvent and an intermediate in the manufacture of other organic compounds for the finished products of such as curing agents, engineering plastics andcross-linking agents.
Solubility
It is insoluble in water. But soluble in ethanol, benzene,carbon tetrachloride.
Notes
Stable under ordinary conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
  1. SN Rogak.; U Pott.; T Dann.; D Wang. Gaseous emissions from vehicles in a traffic tunnel in Vancouver, British Columbia. Journal of the Air & Waste Management Association. 1998, 48,(7), 604-615.