2,4-Dichlorobenzyl alcohol, 99%
2,4-Dichlorobenzyl alcohol, 99%
2,4-Dichlorobenzyl alcohol, 99%
2,4-Dichlorobenzyl alcohol, 99%
Thermo Scientific Chemicals
2,4-Dichlorobenzyl alcohol, 99%
CAS: 1777-82-8 | C7H6Cl2O | 177.024 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA13338.22
Price (MYR)
920.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
920.00
EA
Add to cart
Specifications
Chemical Name or Material2,4-Dichlorobenzyl alcohol
CAS1777-82-8
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H318
Causes serious eye damage.
Health Hazard 3GHS P Statement
P280i-P305+P351+P338-P337+P313
Wear eye/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.
View more
2,4-Dichlorobenzyl alcohol is an antiseptic found in commercially available throat lozenges. Also used deactivate Respiratory syncytial virus and SARS-Cov.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4-Dichlorobenzyl alcohol is an antiseptic found in commercially available throat lozenges. Also used deactivate Respiratory syncytial virus and SARS-Cov.
Solubility
Soluble in water, methanol and chloroform.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Alex Thompson; Sandie Reader; Emma Field; Adrian Shephard. Open-label taste-testing study to evaluate the acceptability of both strawberry-flavored and orange-flavored amylmetacresol/2,4-dichlorobenzyl alcohol throat lozenges in healthy children. Drugs in R&D,. 2013, 13 (2), 101-107.
  2. I Abraham; R L Dion; D M Chi; M M Gottesman; and E Hamel. 2,4-Dichlorobenzyl thiocyanate, an antimitotic agent that alters microtubule morphology. 1986, 83 (11), 6839-6843.