2-Isopropylimidazole, 98%
2-Isopropylimidazole, 98%
2-Isopropylimidazole, 98%
2-Isopropylimidazole, 98%
Thermo Scientific Chemicals
2-Isopropylimidazole, 98%
CAS: 36947-68-9 | C6H10N2 | 110.16 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13307.22100 g
Catalog number ALFA13307.22
Price (MYR)
378.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
378.00
EA
Add to cart
Specifications
Chemical Name or Material2-Isopropylimidazole
CAS36947-68-9
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c